1,2,3,9b-Tetrahydro-4λ4-benzo[c]thieno[2,1-e]isothiazole-8-carbonitrile 4-oxide 95%

Lieferant: Apollo Scientific
Gefahr

OR55428-1G OR55428-250MG
APOSOR55428-1GEA 1010 EUR
APOSOR55428-1G APOSOR55428-250MG
1,2,3,9b-Tetrahydro-4λ4-benzo[c]thieno[2,1-e]isothiazole-8-carbonitrile 4-oxide 95%
1,2,3,9b-Tetrahydro-4λ4-benzo[c]thieno[2,1-e]isothiazole-8-carbonitrile 4-oxide
Formel: C₁₁H₁₀N₂OS
Molecular Weight: 218.275 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31629909
CAS-Nummer: 2169310-98-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR