6-Amino-3-bromo-2-methoxybenzonitrile

Lieferant: Apollo Scientific

OR55462-1G OR55462-500MG
APOSOR55462-1GEA 691 EUR
APOSOR55462-1G APOSOR55462-500MG
6-Amino-3-bromo-2-methoxybenzonitrile
6-Amino-3-bromo-2-methoxybenzonitrile
Formel: C₈H₇BrN₂O
Molecular Weight: 227,058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23115406
CAS-Nummer: 1421312-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR