Methyl amino(2-chlorophenyl)acetate hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-amino-2-(2-chlorophenyl)acetate hydrochloride

OR55467-1G OR55467-5G
APOSOR55467-1GEA 33.5 EUR
APOSOR55467-1G APOSOR55467-5G
Methyl amino(2-chlorophenyl)acetate hydrochloride 95%
Methyl amino(2-chlorophenyl)acetate hydrochloride
Formel: C₉H₁₀ClNO₂·ClH
Molecular Weight: 236.095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16661100
CAS-Nummer: 141109-17-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR