2-Chloro-6-(methylamino)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methylamino-6-chlorobenzoic acid

OR55471-1G OR55471-5G
APOSOR55471-1GEA 437 EUR
APOSOR55471-1G APOSOR55471-5G
2-Chloro-6-(methylamino)benzoic acid 97%
2-Chloro-6-(methylamino)benzoic acid
Formel: C₈H₈ClNO₂
Molecular Weight: 185,608 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16852535
CAS-Nummer: 64460-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR