(S)-4-Methyl-2-(hydroxymethyl)morpholine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-(4-Methylmorpholin-2-yl)methanol

OR55501-1G OR55501-250MG
APOSOR55501-1GEA 301 EUR
APOSOR55501-1G APOSOR55501-250MG
(S)-4-Methyl-2-(hydroxymethyl)morpholine 95%
(S)-4-Methyl-2-(hydroxymethyl)morpholine
Formel: C₆H₁₃NO₂
Molecular Weight: 131,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16294695
CAS-Nummer: 1159598-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR