3-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, pinacol ester 96%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

OR5552-1G OR5552-5G
APOSOR5552-1GEA 162 EUR
APOSOR5552-1G APOSOR5552-5G
3-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, pinacol ester 96%
3-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, pinacol ester
Formel: C₁₅H₂₀BClO₄
Molecular Weight: 310.581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659929
CAS-Nummer: 474709-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR