2-Bromo-6-hydroxybenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-2-(hydroxymethyl)phenol

OR55558-1G OR55558-500MG OR55558-5G
APOSOR55558-1GEA 267 EUR
APOSOR55558-1G APOSOR55558-500MG APOSOR55558-5G
2-Bromo-6-hydroxybenzyl alcohol
2-Bromo-6-hydroxybenzyl alcohol
Formel: C₇H₇BrO₂
Molecular Weight: 203.033 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18391434
CAS-Nummer: 96911-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR