5-Chloro-1-methyl-1H-pyrazole-4-carbaldehyde 95%
Lieferant: Apollo Scientific
OR55584-100MG
OR55584-250MG
APOSOR55584-100MGEA
48
EUR
APOSOR55584-100MG
APOSOR55584-250MG
5-Chloro-1-methyl-1H-pyrazole-4-carbaldehyde 95%
5-Chloro-1-methyl-1H-pyrazole-4-carbaldehyde
Formel:
C₅H₅ClN₂O Molecular Weight: 144.559 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16619816 CAS-Nummer: 117007-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...