4-Chloro-5-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR55604-1G OR55604-250MG
APOSOR55604-1GEA 917 EUR
APOSOR55604-1G APOSOR55604-250MG
4-Chloro-5-methyl-1H-indazole
4-Chloro-5-methyl-1H-indazole
Formel: C₈H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22628251
CAS-Nummer: 152626-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR