2-(Bromomethyl)aniline, N-BOC protected 96%
Lieferant: Apollo Scientific
Gefahr
Synonyme: tert-Butyl (2-(bromomethyl)phenyl)carbamate
OR55620-1G OR55620-5G
APOSOR55620-1GEA 359 EUR
APOSOR55620-1G APOSOR55620-5G
2-(Bromomethyl)aniline, N-BOC protected 96%
2-(Bromomethyl)aniline, N-BOC protected
Formel: C₁₂H₁₆BrNO₂
Molecular Weight: 286,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11616720
CAS-Nummer: 166329-43-7
UN: 3261
ADR: 8,III