2-Bromo-5-methoxybenzenesulphonamide 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR55625-1G
OR55625-250MG
APOSOR55625-1GEA
1510
EUR
APOSOR55625-1G
APOSOR55625-250MG
2-Bromo-5-methoxybenzenesulphonamide 95%
2-Bromo-5-methoxybenzenesulphonamide
Formel:
C₇H₈BrNO₃S Molecular Weight: 266,112 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18379741 CAS-Nummer: 749253-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...