Ethyl 4-bromo-1,3-thiazole-2-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4-bromothiazole-2-carboxylate

OR55633-1G OR55633-5G
APOSOR55633-1GEA 53.8 EUR
APOSOR55633-1G APOSOR55633-5G
Ethyl 4-bromo-1,3-thiazole-2-carboxylate 95%
Ethyl 4-bromo-1,3-thiazole-2-carboxylate
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,086 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11044971
CAS-Nummer: 959755-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR