7-Bromo-5-chlorobenzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR55635-1G OR55635-250MG
APOSOR55635-1GEA 77 EUR
APOSOR55635-1G APOSOR55635-250MG
7-Bromo-5-chlorobenzofuran-2-carboxylic acid
7-Bromo-5-chlorobenzofuran-2-carboxylic acid
Formel: C₉H₄BrClO₃
Molecular Weight: 275,483 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04208738
CAS-Nummer: 190775-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR