5-Isocyanato-1-methyl-1H-indole 97%

Lieferant: Apollo Scientific
Gefahr

OR55642-1G OR55642-250MG
APOSOR55642-1GEA 190 EUR
APOSOR55642-1G APOSOR55642-250MG
5-Isocyanato-1-methyl-1H-indole 97%
5-Isocyanato-1-methyl-1H-indole

When melted, may contain small amount of sediment

Formel: C₁₀H₈N₂O
Molecular Weight: 172,183 g/mol
Schmelzpunkt: 42…45 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09702358
CAS-Nummer: 884507-16-8
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR