Methyl pent-4-ynoate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Pentynoic acid methyl ester
OR55646-1G
OR55646-250MG
OR55646-5G
OR55646-25G
APOSOR55646-1GEA
21.9
EUR
APOSOR55646-1G
APOSOR55646-250MG
APOSOR55646-5G
APOSOR55646-25G
Methyl pent-4-ynoate
Methyl pent-4-ynoate
Formel:
C₆H₈O₂ Molecular Weight: 112,127 g/mol Siedepunkt: 141…142 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD12025043 CAS-Nummer: 21565-82-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...