4-Hydroxy-5-methyl-7-nitro-2,3-dihydro-1H-indene 97%

Lieferant: Apollo Scientific
Achtung

OR55648-1G OR55648-250MG
APOSOR55648-1GEA 944 EUR
APOSOR55648-1G APOSOR55648-250MG
4-Hydroxy-5-methyl-7-nitro-2,3-dihydro-1H-indene 97%
4-Hydroxy-5-methyl-7-nitro-2,3-dihydro-1H-indene
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,199 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD23133365
CAS-Nummer: 575504-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR