2-[[Dimethyl(oxo)-lambda6-sulphanylidene]amino]acetic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR55657-100MG
OR55657-1G
OR55657-250MG
APOSOR55657-100MGEA
580
EUR
APOSOR55657-100MG
APOSOR55657-1G
APOSOR55657-250MG
2-[[Dimethyl(oxo)-lambda6-sulphanylidene]amino]acetic acid 95%
2-[[Dimethyl(oxo)-lambda6-sulphanylidene]amino]acetic acid
Formel:
C₄H₉NO₃S Molecular Weight: 151,184 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28397370 CAS-Nummer: 1621962-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...