7-Bromo-5-nitrobenzofuran

Lieferant: Apollo Scientific
Achtung

OR55661-1G OR55661-250MG OR55661-500MG
APOSOR55661-1GEA 209 EUR
APOSOR55661-1G APOSOR55661-250MG APOSOR55661-500MG
7-Bromo-5-nitrobenzofuran
7-Bromo-5-nitrobenzofuran
Formel: C₈H₄BrNO₃
Molecular Weight: 242,026 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28098935
CAS-Nummer: 286836-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR