2-Bromo-4-cyclopropylphenol

Lieferant: Apollo Scientific
Achtung

OR55664-1G OR55664-250MG
APOSOR55664-1GEA 178 EUR
APOSOR55664-1G APOSOR55664-250MG
2-Bromo-4-cyclopropylphenol
2-Bromo-4-cyclopropylphenol
Formel: C₉H₉BrO
Molecular Weight: 213.071 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD31700417
CAS-Nummer: 2386858-57-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR