4-Cyclopropylbenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR55667-1G
OR55667-250MG
APOSOR55667-1GEA
377
EUR
APOSOR55667-1G
APOSOR55667-250MG
4-Cyclopropylbenzonitrile
4-Cyclopropylbenzonitrile
Formel:
C₁₀H₉N Lagertemperatur: Gefrierschrank |
MDL:
MFCD06802400 CAS-Nummer: 1126-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...