2-Bromo-3'-cyclopropylacetophenone

Lieferant: Apollo Scientific
Gefahr

OR55673-1G OR55673-250MG
APOSOR55673-1GEA 109 EUR
APOSOR55673-1G APOSOR55673-250MG
2-Bromo-3'-cyclopropylacetophenone
2-Bromo-3'-cyclopropylacetophenone
Formel: C₁₁H₁₁BrO
Molecular Weight: 239.108 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD20484207
CAS-Nummer: 627463-23-4
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR