Ethyl 3-amino-6-bromobenzofuran-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR55680-1G
OR55680-250MG
APOSOR55680-1GEA
63.8
EUR
APOSOR55680-1G
APOSOR55680-250MG
Ethyl 3-amino-6-bromobenzofuran-2-carboxylate
Ethyl 3-amino-6-bromobenzofuran-2-carboxylate
Formel:
C₁₁H₁₀BrNO₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD21609331 CAS-Nummer: 887250-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...