3-Iodo-2-methylbenzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(Bromomethyl)-2-methyliodobenzene
OR55687-1G
OR55687-5G
APOSOR55687-1GEA
94.4
EUR
APOSOR55687-1G
APOSOR55687-5G
3-Iodo-2-methylbenzyl bromide 95%
3-Iodo-2-methylbenzyl bromide
Formel:
C₈H₈BrI Molecular Weight: 310,958 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18397827 CAS-Nummer: 950577-16-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...