Ethyl 1-phenyl-1,2,4-triazole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR55689-1G OR55689-250MG
APOSOR55689-1GEA 221 EUR
APOSOR55689-1G APOSOR55689-250MG
Ethyl 1-phenyl-1,2,4-triazole-3-carboxylate
Ethyl 1-phenyl-1,2,4-triazole-3-carboxylate
Formel: C₁₁H₁₁N₃O₂
Molecular Weight: 217,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03837854
CAS-Nummer: 1019-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR