1-(4-Bromophenyl)-1,2,4-triazole-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR55692-1G OR55692-250MG
APOSOR55692-1GEA 121 EUR
APOSOR55692-1G APOSOR55692-250MG
1-(4-Bromophenyl)-1,2,4-triazole-3-carboxylic acid
1-(4-Bromophenyl)-1,2,4-triazole-3-carboxylic acid
Formel: C₉H₆BrN₃O₂
Molecular Weight: 268.067 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29765140
CAS-Nummer: 1782518-89-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR