4-Bromo-1-benzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromobenzofuran-2-carboxylic acid

OR55703-1G OR55703-250MG OR55703-5G
APOSOR55703-1GEA 169 EUR
APOSOR55703-1G APOSOR55703-250MG APOSOR55703-5G
4-Bromo-1-benzofuran-2-carboxylic acid
4-Bromo-1-benzofuran-2-carboxylic acid
Formel: C₉H₅BrO₃
Molecular Weight: 241.038 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22034498
CAS-Nummer: 1369143-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR