Ethyl 7-bromobenzofuran-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR55717-1G OR55717-250MG OR55717-5G
APOSOR55717-1GEA 252 EUR
APOSOR55717-1G APOSOR55717-250MG APOSOR55717-5G
Ethyl 7-bromobenzofuran-2-carboxylate
Ethyl 7-bromobenzofuran-2-carboxylate
Formel: C₁₁H₉BrO₃
Molecular Weight: 269,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699643
CAS-Nummer: 1033201-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR