4-Bromo-1,3-dimethyl-1H-pyrazole-5-carbonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
OR55728-1G
OR55728-250MG
APOSOR55728-1GEA
232
EUR
APOSOR55728-1G
APOSOR55728-250MG
4-Bromo-1,3-dimethyl-1H-pyrazole-5-carbonitrile
4-Bromo-1,3-dimethyl-1H-pyrazole-5-carbonitrile
Formel:
C₆H₆BrN₃ Molecular Weight: 200,036 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30491859 CAS-Nummer: 1454852-95-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...