3-Iodo-4-methylbenzaldehyde

Lieferant: Apollo Scientific
Achtung

OR55732-1G OR55732-250MG
APOSOR55732-1GEA 219 EUR
APOSOR55732-1G APOSOR55732-250MG
3-Iodo-4-methylbenzaldehyde
3-Iodo-4-methylbenzaldehyde
Formel: C₈H₇IO
Molecular Weight: 246,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08703356
CAS-Nummer: 58586-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR