Ethyl 7-chlorobenzofuran-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR55736-1G OR55736-250MG
APOSOR55736-1GEA 522 EUR
APOSOR55736-1G APOSOR55736-250MG
Ethyl 7-chlorobenzofuran-2-carboxylate
Ethyl 7-chlorobenzofuran-2-carboxylate
Formel: C₁₁H₉ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31745692
CAS-Nummer: 260273-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR