7-Chlorobenzofuran-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR55737-1G
OR55737-250MG
APOSOR55737-1GEA
261
EUR
APOSOR55737-1G
APOSOR55737-250MG
7-Chlorobenzofuran-2-carboxylic acid
7-Chlorobenzofuran-2-carboxylic acid
Formel:
C₉H₅ClO₃ Molecular Weight: 196,587 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060515 CAS-Nummer: 63558-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...