(5-Chlorobenzofuran-2-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
OR55739-1G
OR55739-250MG
APOSOR55739-1GEA
168
EUR
APOSOR55739-1G
APOSOR55739-250MG
(5-Chlorobenzofuran-2-yl)methanol
(5-Chlorobenzofuran-2-yl)methanol
Formel:
C₉H₇ClO₂ Molecular Weight: 182.604 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16877417 CAS-Nummer: 235082-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...