Tris(N,N-dimethylaminopropyl)amine

Lieferant: Apollo Scientific
Gefahr

Synonyme: N1,N1-Bis(3-(Dimethylamino)propyl)-N3,N3-dimethylpropane-1,3-diamine

OR55744-1G OR55744-250MG
APOSOR55744-1GEA 150 EUR
APOSOR55744-1G APOSOR55744-250MG
Tris(N,N-dimethylaminopropyl)amine
Tris(N,N-dimethylaminopropyl)amine
Formel: C₁₅H₃₆N₄
Molecular Weight: 272,473 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00190091
CAS-Nummer: 33329-35-0
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR