4-Hydrazino-1-methylpiperidine dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-Methylpiperidin-4-yl)hydrazine dihydrochloride, 4-Hydrazinyl-1-methylpiperidine dihydrochloride

OR5644-1G OR5644-250MG OR5644-5G
APOSOR5644-1GEA 105 EUR
APOSOR5644-1G APOSOR5644-250MG APOSOR5644-5G
4-Hydrazino-1-methylpiperidine dihydrochloride
4-Hydrazino-1-methylpiperidine dihydrochloride
Formel: C₆H₁₅N₃·2ClH
Molecular Weight: 202.125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059574
CAS-Nummer: 53242-78-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR