2-(Aminomethyl)-1H-indole methanesulphonate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1H-Indol-2-yl)methylamine methanesulphonate, 2-(Aminomethyl)-1H-indole mesylate

OR5660-250MG OR5660-1G
APOSOR5660-250MGEA 372 EUR
APOSOR5660-250MG APOSOR5660-1G
2-(Aminomethyl)-1H-indole methanesulphonate 95%
2-(Aminomethyl)-1H-indole methanesulphonate
Formel: C₉H₁₀N₂·CH₃SO₃H
Molecular Weight: 242.295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03756929
CAS-Nummer: 1072806-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR