1-[(Pyridin-2-yl)methyl]piperidine-3-carboxylic acid dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-[(3-Carboxypiperidin-1-yl)methyl]pyridine dihydrochloride, 3-Carboxy-1-[(pyridin-2-yl)methyl]piperidine dihydrochloride
OR5683-1G
OR5683-250MG
APOSOR5683-1GEA
551
EUR
APOSOR5683-1G
APOSOR5683-250MG
1-[(Pyridin-2-yl)methyl]piperidine-3-carboxylic acid dihydrochloride
1-[(Pyridin-2-yl)methyl]piperidine-3-carboxylic acid dihydrochloride
Formel:
C₁₂H₁₆N₂O₂·2ClH Molecular Weight: 293,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06801257 CAS-Nummer: 1171204-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...