5-(Methoxycarbonyl)pyridine-3-boronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-borononicotinate, Methyl 5-boronopyridine-3-carboxylate, (5-(Methoxycarbonyl)pyridin-3-yl)boronic acid

OR5718-100MG OR5718-250MG OR5718-1G
APOSOR5718-100MGEA 40.6 EUR
APOSOR5718-100MG APOSOR5718-250MG APOSOR5718-1G
5-(Methoxycarbonyl)pyridine-3-boronic acid 97%
5-(Methoxycarbonyl)pyridine-3-boronic acid
Formel: C₇H₈BNO₄
Molecular Weight: 180.954 g/mol
Schmelzpunkt: 102…108 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07363742
CAS-Nummer: 871329-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR