4-Amino-3-hydroxypyridine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Hydroxypyridin-4-amine hydrochloride, 4-Aminopyridin-3-ol hydrochloride

OR5796-250MG OR5796-1G
APOSOR5796-250MGEA 87 EUR
APOSOR5796-250MG APOSOR5796-1G
4-Amino-3-hydroxypyridine hydrochloride
4-Amino-3-hydroxypyridine hydrochloride
Formel: C₅H₆N₂O·ClH
Molecular Weight: 146,575 g/mol
Schmelzpunkt: 222…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08729308
CAS-Nummer: 1206679-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR