Dimethyl 3,3'-bipyridine-5,5'-dicarboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,5'-Bis(methoxycarbonyl)-3,3'-bipyridine

OR5803-1G OR5803-250MG
APOSOR5803-1GEA 869 EUR
APOSOR5803-1G APOSOR5803-250MG
Dimethyl 3,3'-bipyridine-5,5'-dicarboxylate 97%
Dimethyl 3,3'-bipyridine-5,5'-dicarboxylate
Formel: C₁₄H₁₂N₂O₄
Molecular Weight: 272,256 g/mol
Schmelzpunkt: 226…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363810
CAS-Nummer: 128612-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR