5-Acetamido-2-bromopyridine 95%

Lieferant: Apollo Scientific
Achtung

OR5807-1G OR5807-5G
APOSOR5807-1GEA 87 EUR
APOSOR5807-1G APOSOR5807-5G
5-Acetamido-2-bromopyridine 95%
5-Acetamido-2-bromopyridine
Formel: C₇H₇BrN₂O
Molecular Weight: 215,047 g/mol
Schmelzpunkt: 134…140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07363807
CAS-Nummer: 29958-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR