1-[(4-Bromophenyl)sulphonyl]azetidine 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Azetidin-1-yl 4-bromophenyl sulphone, 1-[(Azetidin-1-yl)sulphonyl]-4-bromobenzene, 1-[(4-Bromophenyl)sulphonyl]azetane, 1-((4-Bromophenyl)sulphonyl)azetidine
OR5812-1G
OR5812-5G
APOSOR5812-1GEA
139
EUR
APOSOR5812-1G
APOSOR5812-5G
1-[(4-Bromophenyl)sulphonyl]azetidine 96%
1-[(4-Bromophenyl)sulphonyl]azetidine
Formel:
C₉H₁₀BrNO₂S Molecular Weight: 276,15 g/mol Schmelzpunkt: 118…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07363814 CAS-Nummer: 530081-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...