4-[(Dimethylamino)methyl]piperidine

Lieferant: Apollo Scientific
Achtung

Synonyme: N,N-Dimethyl-1-(piperidin-4-yl)methylamine, N,N-Dimethyl-1-(piperidin-4-yl)methanamine

OR5900-1G OR5900-250MG OR5900-5G
APOSOR5900-1GEA 191 EUR
APOSOR5900-1G APOSOR5900-250MG APOSOR5900-5G
4-[(Dimethylamino)methyl]piperidine
4-[(Dimethylamino)methyl]piperidine
Formel: C₈H₁₈N₂
Molecular Weight: 142.242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05215181
CAS-Nummer: 138022-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR