Methyl 4-bromo-3,5-dimethoxybenzoate 95+%

Lieferant: Apollo Scientific
Achtung

OR5931-5G OR5931-1G
APOSOR5931-5GEA 40.6 EUR
APOSOR5931-5G APOSOR5931-1G
Methyl 4-bromo-3,5-dimethoxybenzoate 95+%
Methyl 4-bromo-3,5-dimethoxybenzoate
Formel: C₁₀H₁₁BrO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06628784
CAS-Nummer: 26050-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR