3-Amino-5-methylpyridine hydrochloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Methylpyridin-3-amine hydrochloride

OR59324-1G OR59324-250MG
APOSOR59324-1GEA 176 EUR
APOSOR59324-1G APOSOR59324-250MG
3-Amino-5-methylpyridine hydrochloride
3-Amino-5-methylpyridine hydrochloride
Formel: C₆H₈N₂·ClH
Molecular Weight: 144,602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878433
CAS-Nummer: 957065-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR