4-(2-Aminophenoxy)benzeneboronic acid hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR59345-1G
OR59345-250MG
APOSOR59345-1GEA
343
EUR
APOSOR59345-1G
APOSOR59345-250MG
4-(2-Aminophenoxy)benzeneboronic acid hydrochloride
4-(2-Aminophenoxy)benzeneboronic acid hydrochloride
Formel:
C₁₂H₁₂BNO₃·ClH Molecular Weight: 265.501 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09878349 CAS-Nummer: 957063-10-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...