1-[(4-Methylphenyl)sulphonyl]-1H-imidazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Tosyl-1H-imidazole-4-carboxylic acid

OR59354-1G OR59354-250MG
APOSOR59354-1GEA 0 EUR
APOSOR59354-1G APOSOR59354-250MG
1-[(4-Methylphenyl)sulphonyl]-1H-imidazole-4-carboxylic acid
1-[(4-Methylphenyl)sulphonyl]-1H-imidazole-4-carboxylic acid
Formel: C₁₁H₁₀N₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD09878406
CAS-Nummer: 957063-02-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR