2-Bromo-6-chloro-3-methoxybenzeneboronic acid

Lieferant: Apollo Scientific

Synonyme: (2-Bromo-6-chloro-3-methoxyphenyl)boronic acid, 2-Bromo-6-chloro-3-methoxyphenylboronic acid

OR59359-1G OR59359-5G
APOSOR59359-1GEA 62.5 EUR
APOSOR59359-1G APOSOR59359-5G
2-Bromo-6-chloro-3-methoxybenzeneboronic acid
2-Bromo-6-chloro-3-methoxybenzeneboronic acid
Formel: C₇H₇BBrClO₃
Molecular Weight: 265,297 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878341
CAS-Nummer: 957062-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR