3-[(2-Hydroxyethyl)carbamoyl]benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-{[(2-Hydroxyethyl)amino]carbonyl}benzeneboronic acid, 3-Borono-N-(2-hydroxyethyl)benzamide

OR59387-1G OR59387-250MG OR59387-5G
APOSOR59387-1GEA 232 EUR
APOSOR59387-1G APOSOR59387-250MG APOSOR59387-5G
3-[(2-Hydroxyethyl)carbamoyl]benzeneboronic acid
3-[(2-Hydroxyethyl)carbamoyl]benzeneboronic acid
Formel: C₉H₁₂BNO₄
Molecular Weight: 209,007 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09878355
CAS-Nummer: 955422-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR