Methyl (4-bromo-1H-pyrazol-1-yl)acetate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(4-bromo-1H-pyrazol-1-yl)acetate

OR59391-5G
APOSOR59391-5GEA 139 EUR
APOSOR59391-5G
Methyl (4-bromo-1H-pyrazol-1-yl)acetate
Methyl (4-bromo-1H-pyrazol-1-yl)acetate
Formel: C₆H₇BrN₂O₂
Molecular Weight: 219,036 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878400
CAS-Nummer: 1072944-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR