4-Bromo-3-chlorobenzenesulphonyl chloride 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Bromo-3-chlorobenzene-1-sulphonyl chloride

OR59832-5G
APOSOR59832-5GEA 65.4 EUR
APOSOR59832-5G
4-Bromo-3-chlorobenzenesulphonyl chloride 98%
4-Bromo-3-chlorobenzenesulphonyl chloride
Formel: C₆H₃BrCl₂O₂S
Molecular Weight: 289,962 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973905
CAS-Nummer: 874801-46-4
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR